Assay Details
| Gene Symbol: chat-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001028161.4 | NP_001023332.1 | 5 - 6 | 836 | 96 | |
| NM_001047514.4 | NP_001040979.1 | 2 - 3 | 292 | 96 | ||
| NM_001380191.1 | NP_001368332.1 | 1 - 2 | 112 | 96 | ||
| NR_138505.1 | 4 - 5 | 1001 | 96 | |||
 Back To Top
 Back To TopMore Information
| Set Membership: |  Amplicon length between 86 and 100  3' Most  Recommended Assay  Caenorhabditis elegans  Probe spans exons | 
 Back To Top
 Back To Top
 chat-1
chat-1 Genomic Map
Genomic Map Back To Top
 Back To Top