Assay Details
| Gene Symbol: cif-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_068217.6 | NP_500618.1 | 4 - 5 | 1179 | 126 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

cif-1| Gene Symbol: cif-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_068217.6 | NP_500618.1 | 4 - 5 | 1179 | 126 | |
Set Membership: |
|