Assay Details
| Gene Symbol: miro-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_068219.4 | NP_500620.1 | 4 - 5 | 1047 | 67 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

miro-1| Gene Symbol: miro-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_068219.4 | NP_500620.1 | 4 - 5 | 1047 | 67 | |
Set Membership: |
|