Assay Details
Gene Symbol: Y17G9B.5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_068230.6 | NP_500631.1 | 2 - 3 | 393 | 104 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Y17G9B.5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_068230.6 | NP_500631.1 | 2 - 3 | 393 | 104 |
Set Membership: |
![]() ![]() ![]() ![]() |