Assay Details
| Gene Symbol: tat-2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001028081.3 | NP_001023252.2 | 20 - 21 | 3792 | 80 | |
| NM_001307518.3 | NP_001294447.1 | 21 - 22 | 3897 | 80 | ||
More Information
Set Membership: |
|

tat-2
Genomic Map