Assay Details
| Gene Symbol: tat-2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001028081.3 | NP_001023252.2 | 21 - 22 | 3907 | 65 | |
| NM_001307515.2 | NP_001294444.1 | 21 - 22 | 3911 | 65 | ||
| NM_001307518.3 | NP_001294447.1 | 22 - 23 | 4012 | 65 | ||
More Information
Set Membership: |
|

tat-2
Genomic Map