Assay Details
| Gene Symbol: pes-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001028235.3 | NP_001023406.1 | 1 - 2 | 98 | 72 | |
| GenBank mRNA | Z28377.1 | - | 94 | 72 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

pes-1| Gene Symbol: pes-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001028235.3 | NP_001023406.1 | 1 - 2 | 98 | 72 | |
| GenBank mRNA | Z28377.1 | - | 94 | 72 | ||
Set Membership: |
|