Assay Details
| Gene Symbol: swsn-6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_068400.6 | NP_500801.2 | 5 - 6 | 406 | 86 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

swsn-6| Gene Symbol: swsn-6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_068400.6 | NP_500801.2 | 5 - 6 | 406 | 86 | |
Set Membership: |
|