Assay Details
| Gene Symbol: spe-27 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_068474.6 | NP_500875.1 | 6 - 7 | 891 | 68 | |
| Gene Symbol: C06E7.88 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001268346.4 | NP_001255275.2 | 6 - 7 | 842 | 68 | |
More Information
Set Membership: |
|

spe-27
Genomic Map