Assay Details
| Gene Symbol: acox-3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_068542.7 | NP_500943.1 | 10 - 11 | 1910 | 128 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

acox-3| Gene Symbol: acox-3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_068542.7 | NP_500943.1 | 10 - 11 | 1910 | 128 | |
Set Membership: |
|