Assay Details
| Gene Symbol: csp-2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001380264.1 | NP_001368632.1 | 7 - 8 | 1357 | 82 | |
| NM_001383103.1 | 10 - 11 | 1659 | 82 | |||
| GenBank mRNA | AF088288.1 | - | 10 - 11 | 1656 | 82 | |
More Information
Set Membership: |
|

csp-2
Genomic Map