Assay Details
| Gene Symbol: csp-2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001383103.1 | 2 - 3 | 107 | 72 | ||
| GenBank mRNA | AF088288.1 | - | 2 - 3 | 104 | 72 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

csp-2| Gene Symbol: csp-2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001383103.1 | 2 - 3 | 107 | 72 | ||
| GenBank mRNA | AF088288.1 | - | 2 - 3 | 104 | 72 | |
Set Membership: |
|