Assay Details
| Gene Symbol: fan-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001383106.1 | 6 - 7 | 925 | 67 | ||
| NM_068596.5 | NP_500997.1 | 5 - 6 | 1142 | 67 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001383106.1 transcripts.
Set Membership: |
|

fan-1
Genomic Map