Assay Details
| Gene Symbol: klp-12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001028178.4 | NP_001023349.1 | 9 - 10 | 2715 | 112 | |
| NM_001129324.3 | NP_001122796.1 | 9 - 10 | 2715 | 112 | ||
| NM_001372925.2 | NP_001360493.1 | 9 - 10 | 2721 | 112 | ||
| Gene Symbol: T01G1.4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001028180.7 | NP_001023351.1 | 15 - 16 | 4041 | 112 | |
More Information
Set Membership: |
|

klp-12
Genomic Map