Assay Details
| Gene Symbol: klp-12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001028178.4 | NP_001023349.1 | 4 - 5 | 932 | 69 | |
| NM_001129324.3 | NP_001122796.1 | 4 - 5 | 932 | 69 | ||
| NM_001372925.2 | NP_001360493.1 | 4 - 5 | 938 | 69 | ||
| Gene Symbol: T01G1.4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001028180.7 | NP_001023351.1 | 10 - 11 | 2250 | 69 | |
More Information
Set Membership: |
|

klp-12
Genomic Map