Assay Details
| Gene Symbol: kin-4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001268622.2 | NP_001255551.1 | 17 - 18 | 3680 | 83 | |
| NM_001268624.3 | NP_001255553.1 | 17 - 18 | 3671 | 83 | ||
| NM_001268625.3 | NP_001255554.1 | 16 - 17 | 3134 | 83 | ||
| NM_001268626.3 | NP_001255555.1 | 15 - 16 | 3122 | 83 | ||
| NM_001268627.2 | NP_001255556.1 | 17 - 18 | 3671 | 83 | ||
| NM_001268629.3 | NP_001255558.1 | 12 - 13 | 2333 | 83 | ||
| NM_001307048.3 | NP_001293977.1 | 14 - 15 | 2891 | 83 | ||
| NM_001380461.1 | NP_001367231.1 | 16 - 17 | 3575 | 83 | ||
| NM_001380462.2 | NP_001366913.1 | 16 - 17 | 3614 | 83 | ||
| GenBank mRNA | AM086627.1 | - | 14 - 15 | 2882 | 83 | |
More Information
Set Membership: |
|

kin-4
Genomic Map