Assay Details
| Gene Symbol: kin-4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001268622.2 | NP_001255551.1 | 19 - 20 | 4913 | 85 | |
| NM_001268624.3 | NP_001255553.1 | 19 - 20 | 4904 | 85 | ||
| NM_001268625.3 | NP_001255554.1 | 18 - 19 | 4367 | 85 | ||
| NM_001268626.3 | NP_001255555.1 | 17 - 18 | 4355 | 85 | ||
| NM_001268629.3 | NP_001255558.1 | 14 - 15 | 3566 | 85 | ||
| NM_001307048.3 | NP_001293977.1 | 16 - 17 | 4124 | 85 | ||
| NM_001380461.1 | NP_001367231.1 | 18 - 19 | 4808 | 85 | ||
| NM_001380462.2 | NP_001366913.1 | 18 - 19 | 4847 | 85 | ||
| GenBank mRNA | AM086627.1 | - | 16 - 17 | 4115 | 85 | |
More Information
Set Membership: |
|

kin-4
Genomic Map