Assay Details
| Gene Symbol: kin-4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001268622.2 | NP_001255551.1 | 20 - 21 | 5176 | 69 | |
| NM_001268624.3 | NP_001255553.1 | 20 - 21 | 5167 | 69 | ||
| NM_001268625.3 | NP_001255554.1 | 19 - 20 | 4630 | 69 | ||
| NM_001268626.3 | NP_001255555.1 | 18 - 19 | 4618 | 69 | ||
| NM_001268629.3 | NP_001255558.1 | 15 - 16 | 3829 | 69 | ||
| NM_001307048.3 | NP_001293977.1 | 17 - 18 | 4387 | 69 | ||
| NM_001380461.1 | NP_001367231.1 | 19 - 20 | 5071 | 69 | ||
| NM_001380462.2 | NP_001366913.1 | 19 - 20 | 5110 | 69 | ||
| GenBank mRNA | AM086627.1 | - | 17 - 18 | 4378 | 69 | |
More Information
Set Membership: |
|

kin-4
Genomic Map