Assay Details
| Gene Symbol: kin-4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001268622.2 | NP_001255551.1 | 8 - 9 | 1901 | 122 | |
| NM_001268624.3 | NP_001255553.1 | 8 - 9 | 1892 | 122 | ||
| NM_001268625.3 | NP_001255554.1 | 7 - 8 | 1355 | 122 | ||
| NM_001268626.3 | NP_001255555.1 | 6 - 7 | 1343 | 122 | ||
| NM_001268627.2 | NP_001255556.1 | 8 - 9 | 1892 | 122 | ||
| NM_001268629.3 | NP_001255558.1 | 3 - 4 | 554 | 122 | ||
| NM_001307048.3 | NP_001293977.1 | 5 - 6 | 1112 | 122 | ||
| NM_001380461.1 | NP_001367231.1 | 7 - 8 | 1796 | 122 | ||
| NM_001380462.2 | NP_001366913.1 | 7 - 8 | 1835 | 122 | ||
| GenBank mRNA | AM086627.1 | - | 5 - 6 | 1103 | 122 | |
More Information
Set Membership: |
|

kin-4
Genomic Map