Assay Details
| Gene Symbol: kin-4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001268622.2 | NP_001255551.1 | 14 - 15 | 3129 | 101 | |
| NM_001268624.3 | NP_001255553.1 | 14 - 15 | 3120 | 101 | ||
| NM_001268625.3 | NP_001255554.1 | 13 - 14 | 2583 | 101 | ||
| NM_001268626.3 | NP_001255555.1 | 12 - 13 | 2571 | 101 | ||
| NM_001268627.2 | NP_001255556.1 | 14 - 15 | 3120 | 101 | ||
| NM_001268629.3 | NP_001255558.1 | 9 - 10 | 1782 | 101 | ||
| NM_001307048.3 | NP_001293977.1 | 11 - 12 | 2340 | 101 | ||
| NM_001380461.1 | NP_001367231.1 | 13 - 14 | 3024 | 101 | ||
| NM_001380462.2 | NP_001366913.1 | 13 - 14 | 3063 | 101 | ||
| GenBank mRNA | AM086627.1 | - | 11 - 12 | 2331 | 101 | |
More Information
Set Membership: |
|

kin-4
Genomic Map