Assay Details
| Gene Symbol: mep-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_069772.8 | NP_502173.2 | 1 - 2 | 120 | 72 | |
| GenBank mRNA | AF416567.1 | - | 1 - 2 | 142 | 72 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

mep-1| Gene Symbol: mep-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_069772.8 | NP_502173.2 | 1 - 2 | 120 | 72 | |
| GenBank mRNA | AF416567.1 | - | 1 - 2 | 142 | 72 | |
Set Membership: |
|