Assay Details
| Gene Symbol: mep-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_069772.8 | NP_502173.2 | 3 - 4 | 1409 | 73 | |
| GenBank mRNA | AF416567.1 | - | 3 - 4 | 1431 | 73 | |
More Information
Set Membership: |
|

mep-1
Genomic Map