Assay Details
| Gene Symbol: rib-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_069779.5 | NP_502180.1 | 4 - 5 | 488 | 86 | |
| GenBank mRNA | AB241458.1 | - | 4 - 5 | 523 | 86 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

rib-1| Gene Symbol: rib-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_069779.5 | NP_502180.1 | 4 - 5 | 488 | 86 | |
| GenBank mRNA | AB241458.1 | - | 4 - 5 | 523 | 86 | |
Set Membership: |
|