Assay Details
| Gene Symbol: rib-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_069779.5 | NP_502180.1 | 6 - 7 | 732 | 124 | |
| GenBank mRNA | AB241458.1 | - | 6 - 7 | 767 | 124 | |
More Information
Set Membership: |
|

rib-1
Genomic Map