Assay Details
| Gene Symbol: mars-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001268641.3 | NP_001255570.1 | 5 - 6 | 2640 | 69 | |
| NM_001268642.3 | NP_001255571.1 | 2 - 3 | 754 | 69 | ||
More Information
Set Membership: |
|

mars-1
Genomic Map