Assay Details
| Gene Symbol: dpy-20 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_069814.6 | NP_502215.2 | 4 - 5 | 1620 | 102 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

dpy-20| Gene Symbol: dpy-20 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_069814.6 | NP_502215.2 | 4 - 5 | 1620 | 102 | |
Set Membership: |
|