Assay Details
| Gene Symbol: magi-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001268648.3 | NP_001255577.1 | 10 - 11 | 1407 | 82 | |
| NM_001268650.4 | NP_001255579.1 | 5 - 6 | 997 | 82 | ||
| NM_001268652.3 | NP_001255581.1 | 3 - 4 | 681 | 82 | ||
| NM_001383205.1 | 10 - 11 | 1397 | 82 | |||
| NM_001383206.1 | 10 - 11 | 1413 | 82 | |||
More Information
Set Membership: |
|

magi-1
Genomic Map