Assay Details
| Gene Symbol: gem-4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_069824.4 | NP_502225.1 | 6 - 7 | 1416 | 126 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

gem-4| Gene Symbol: gem-4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_069824.4 | NP_502225.1 | 6 - 7 | 1416 | 126 | |
Set Membership: |
|