Assay Details
| Gene Symbol: par-5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001380472.1 | NP_001368180.1 | 1 - 2 | 86 | 66 | |
| NM_001383210.1 | 3 - 4 | 479 | 66 | |||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

par-5| Gene Symbol: par-5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001380472.1 | NP_001368180.1 | 1 - 2 | 86 | 66 | |
| NM_001383210.1 | 3 - 4 | 479 | 66 | |||
Set Membership: |
|