Assay Details
| Gene Symbol: ech-2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001268663.2 | NP_001255592.1 | 1 - 2 | 85 | 87 | |
| NM_001268664.3 | NP_001255593.1 | 1 - 2 | 87 | 87 | ||
| Gene Symbol: tag-30 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001380473.1 | NP_001366838.1 | 1 - 2 | 115 | 87 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001380473.1 transcripts.
Set Membership: |
|

ech-2
Genomic Map