Assay Details
| Gene Symbol: ech-2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001268663.2 | NP_001255592.1 | 3 - 4 | 261 | 81 | |
| NM_001268664.3 | NP_001255593.1 | 2 - 3 | 305 | 81 | ||
| NR_131613.1 | 3 - 4 | 260 | 81 | |||
| Gene Symbol: tag-30 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001380473.1 | NP_001366838.1 | 3 - 4 | 291 | 81 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001380473.1 transcripts.
Set Membership: |
|

ech-2
Genomic Map