Assay Details
| Gene Symbol: trpa-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001307154.4 | NP_001294083.1 | 2 - 3 | 515 | 115 | |
| NM_069848.7 | NP_502249.3 | 2 - 3 | 512 | 115 | ||
More Information
Set Membership: |
|

trpa-1
Genomic Map