Assay Details
| Gene Symbol: trpa-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001307154.4 | NP_001294083.1 | 6 - 7 | 1844 | 102 | |
| NM_069848.7 | NP_502249.3 | 6 - 7 | 1841 | 102 | ||
More Information
Set Membership: |
|

trpa-1
Genomic Map