Assay Details
| Gene Symbol: unc-22 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001129363.4 | NP_001122835.2 | 8 - 9 | 1539 | 85 | |
| NM_001268672.3 | NP_001255601.1 | 14 - 15 | 2202 | 85 | ||
| NM_001268673.3 | NP_001255602.1 | 14 - 15 | 1938 | 85 | ||
| NM_001268674.4 | NP_001255603.2 | 5 - 6 | 777 | 85 | ||
| NM_001268675.3 | NP_001255604.1 | 4 - 5 | 726 | 85 | ||
| NM_001307045.3 | NP_001293974.1 | 16 - 17 | 2820 | 85 | ||
| NM_001307046.3 | NP_001293975.1 | 16 - 17 | 2820 | 85 | ||
| NM_069872.6 | NP_502273.2 | 14 - 15 | 2065 | 85 | ||
| NM_069873.6 | NP_502274.2 | 17 - 18 | 2895 | 85 | ||
More Information
Set Membership: |
|

unc-22
Genomic Map