Assay Details
| Gene Symbol: unc-22 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001129363.4 | NP_001122835.2 | 9 - 10 | 1624 | 65 | |
| NM_001268672.3 | NP_001255601.1 | 15 - 16 | 2287 | 65 | ||
| NM_001268673.3 | NP_001255602.1 | 15 - 16 | 2023 | 65 | ||
| NM_001268674.4 | NP_001255603.2 | 6 - 7 | 862 | 65 | ||
| NM_001268675.3 | NP_001255604.1 | 5 - 6 | 811 | 65 | ||
| NM_001307045.3 | NP_001293974.1 | 17 - 18 | 2905 | 65 | ||
| NM_001307046.3 | NP_001293975.1 | 17 - 18 | 2905 | 65 | ||
| NM_069872.6 | NP_502273.2 | 15 - 16 | 2150 | 65 | ||
| NM_069873.6 | NP_502274.2 | 18 - 19 | 2980 | 65 | ||
More Information
Set Membership: |
|

unc-22
Genomic Map