Assay Details
| Gene Symbol: unc-22 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001129363.4 | NP_001122835.2 | 10 - 11 | 1791 | 74 | |
| NM_001268672.3 | NP_001255601.1 | 16 - 17 | 2454 | 74 | ||
| NM_001268673.3 | NP_001255602.1 | 16 - 17 | 2190 | 74 | ||
| NM_001268674.4 | NP_001255603.2 | 7 - 8 | 1029 | 74 | ||
| NM_001268675.3 | NP_001255604.1 | 6 - 7 | 978 | 74 | ||
| NM_001307045.3 | NP_001293974.1 | 18 - 19 | 3072 | 74 | ||
| NM_001307046.3 | NP_001293975.1 | 18 - 19 | 3072 | 74 | ||
| NM_069872.6 | NP_502273.2 | 16 - 17 | 2317 | 74 | ||
| NM_069873.6 | NP_502274.2 | 19 - 20 | 3147 | 74 | ||
More Information
Set Membership: |
|

unc-22
Genomic Map