Assay Details
| Gene Symbol: unc-22 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001129363.4 | NP_001122835.2 | 12 - 13 | 2074 | 109 | |
| NM_001268672.3 | NP_001255601.1 | 18 - 19 | 2737 | 109 | ||
| NM_001268673.3 | NP_001255602.1 | 19 - 20 | 2500 | 109 | ||
| NM_001268674.4 | NP_001255603.2 | 9 - 10 | 1312 | 109 | ||
| NM_001268675.3 | NP_001255604.1 | 8 - 9 | 1261 | 109 | ||
| NM_001307045.3 | NP_001293974.1 | 21 - 22 | 3382 | 109 | ||
| NM_001307046.3 | NP_001293975.1 | 20 - 21 | 3355 | 109 | ||
| NM_069872.6 | NP_502273.2 | 18 - 19 | 2600 | 109 | ||
| NM_069873.6 | NP_502274.2 | 21 - 22 | 3430 | 109 | ||
More Information
Set Membership: |
|

unc-22
Genomic Map