Assay Details
| Gene Symbol: unc-22 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001129363.4 | NP_001122835.2 | 16 - 17 | 7393 | 83 | |
| NM_001268672.3 | NP_001255601.1 | 22 - 23 | 8056 | 83 | ||
| NM_001268673.3 | NP_001255602.1 | 23 - 24 | 7819 | 83 | ||
| NM_001268674.4 | NP_001255603.2 | 13 - 14 | 6631 | 83 | ||
| NM_001268675.3 | NP_001255604.1 | 12 - 13 | 6580 | 83 | ||
| NM_001307045.3 | NP_001293974.1 | 25 - 26 | 8701 | 83 | ||
| NM_001307046.3 | NP_001293975.1 | 24 - 25 | 8674 | 83 | ||
| NM_069872.6 | NP_502273.2 | 22 - 23 | 7919 | 83 | ||
| NM_069873.6 | NP_502274.2 | 25 - 26 | 8749 | 83 | ||
More Information
Set Membership: |
|

unc-22
Genomic Map