Assay Details
| Gene Symbol: unc-22 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001129363.4 | NP_001122835.2 | 21 - 22 | 18054 | 76 | |
| NM_001268672.3 | NP_001255601.1 | 27 - 28 | 18717 | 76 | ||
| NM_001268673.3 | NP_001255602.1 | 28 - 29 | 18480 | 76 | ||
| NM_001268674.4 | NP_001255603.2 | 18 - 19 | 17292 | 76 | ||
| NM_001268675.3 | NP_001255604.1 | 17 - 18 | 17241 | 76 | ||
| NM_001307045.3 | NP_001293974.1 | 30 - 31 | 19362 | 76 | ||
| NM_001307046.3 | NP_001293975.1 | 29 - 30 | 19335 | 76 | ||
| NM_069872.6 | NP_502273.2 | 27 - 28 | 18580 | 76 | ||
| NM_069873.6 | NP_502274.2 | 30 - 31 | 19410 | 76 | ||
More Information
Set Membership: |
|

unc-22
Genomic Map