Assay Details
| Gene Symbol: unc-22 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001129363.4 | NP_001122835.2 | 23 - 24 | 19924 | 67 | |
| NM_001268672.3 | NP_001255601.1 | 29 - 30 | 20587 | 67 | ||
| NM_001268673.3 | NP_001255602.1 | 30 - 31 | 20350 | 67 | ||
| NM_001268674.4 | NP_001255603.2 | 20 - 21 | 19162 | 67 | ||
| NM_001268675.3 | NP_001255604.1 | 19 - 20 | 19111 | 67 | ||
| NM_001307045.3 | NP_001293974.1 | 32 - 33 | 21232 | 67 | ||
| NM_001307046.3 | NP_001293975.1 | 31 - 32 | 21205 | 67 | ||
| NM_069872.6 | NP_502273.2 | 29 - 30 | 20450 | 67 | ||
| NM_069873.6 | NP_502274.2 | 32 - 33 | 21280 | 67 | ||
More Information
Set Membership: |
|

unc-22
Genomic Map