Assay Details
| Gene Symbol: unc-22 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001268672.3 | NP_001255601.1 | 2 - 3 | 209 | 90 | |
| NM_001268673.3 | NP_001255602.1 | 2 - 3 | 209 | 90 | ||
| NM_001307045.3 | NP_001293974.1 | 2 - 3 | 209 | 90 | ||
| NM_001307046.3 | NP_001293975.1 | 2 - 3 | 209 | 90 | ||
| NM_069872.6 | NP_502273.2 | 2 - 3 | 336 | 90 | ||
| NM_069873.6 | NP_502274.2 | 2 - 3 | 209 | 90 | ||
More Information
Set Membership: |
|

unc-22
Genomic Map