Assay Details
| Gene Symbol: mon-2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001268693.2 | NP_001255622.1 | 1 - 2 | 311 | 95 | |
| NM_001268694.2 | NP_001255623.1 | 1 - 2 | 311 | 95 | ||
More Information
Set Membership: |
|

mon-2
Genomic Map