Assay Details
| Gene Symbol: phy-2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_069916.6 | NP_502317.1 | 2 - 3 | 168 | 55 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

phy-2| Gene Symbol: phy-2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_069916.6 | NP_502317.1 | 2 - 3 | 168 | 55 | |
Set Membership: |
|