Assay Details
| Gene Symbol: vps-8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001027888.5 | NP_001023059.1 | 10 - 11 | 3489 | 102 | |
| NM_001307062.3 | NP_001293991.1 | 8 - 9 | 3069 | 102 | ||
More Information
Set Membership: |
|

vps-8
Genomic Map