Assay Details
| Gene Symbol: vps-8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001027888.5 | NP_001023059.1 | 1 - 2 | 103 | 101 | |
| NM_001372932.2 | NP_001359706.1 | 1 - 2 | 101 | 101 | ||
More Information
Set Membership: |
|

vps-8
Genomic Map