Assay Details
| Gene Symbol: vps-8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001027888.5 | NP_001023059.1 | 3 - 4 | 500 | 76 | |
| NM_001307062.3 | NP_001293991.1 | 1 - 2 | 80 | 76 | ||
More Information
Set Membership: |
|

vps-8
Genomic Map