Assay Details
| Gene Symbol: vps-8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001027888.5 | NP_001023059.1 | 5 - 6 | 1909 | 158 | |
| NM_001307062.3 | NP_001293991.1 | 3 - 4 | 1489 | 158 | ||
More Information
Set Membership: |
|

vps-8
Genomic Map