Assay Details
| Gene Symbol: ccr-4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001028436.4 | NP_001023607.1 | 4 - 5 | 707 | 73 | |
| NM_001028437.4 | NP_001023608.1 | 4 - 5 | 711 | 73 | ||
| NM_069954.6 | NP_502355.2 | 4 - 5 | 684 | 73 | ||
| GenBank mRNA | AY043268.1 | - | 4 - 5 | 574 | 73 | |
More Information
Set Membership: |
|

ccr-4
Genomic Map