Assay Details
| Gene Symbol: ccr-4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001028436.4 | NP_001023607.1 | 7 - 8 | 1776 | 113 | |
| NM_001028437.4 | NP_001023608.1 | 7 - 8 | 1780 | 113 | ||
| NM_001136405.2 | NP_001129877.1 | 4 - 5 | 2127 | 113 | ||
| NM_001307061.3 | NP_001293990.1 | 3 - 4 | 1025 | 113 | ||
| NM_069954.6 | NP_502355.2 | 7 - 8 | 1753 | 113 | ||
| GenBank mRNA | AY043268.1 | - | 7 - 8 | 1643 | 113 | |
More Information
Set Membership: |
|

ccr-4
Genomic Map