Assay Details
| Gene Symbol: wdr-46 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_069957.7 | NP_502358.1 | 3 - 4 | 1435 | 101 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

wdr-46| Gene Symbol: wdr-46 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_069957.7 | NP_502358.1 | 3 - 4 | 1435 | 101 | |
Set Membership: |
|